Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PUM2 SIRNA Assays ›
Gene Symbol:
PUM2
Gene Name:
pumilio RNA binding family member 2
Gene Aliases:
PUMH2, PUML2
Chromosome Location:
Chr.2: 20248692 - 20352423 on Build GRCh38
UniGene ID:
Hs.467824
Species:
Human
siRNA ID
HSS118654
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PUM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282752.1
NP_001269681.1
13
2342
NM_001282790.1
NP_001269719.1
12
1968
NM_001282791.1
NP_001269720.1
13
2093
NM_015317.2
NP_056132.1
14
2451
XM_005262607.1
XP_005262664.1
14
2425
XM_005262609.1
XP_005262666.1
13
2188
XM_005262610.1
XP_005262667.1
13
2188
XM_006711973.1
XP_006712036.1
14
2222
XM_006711974.2
XP_006712037.1
14
2262
XM_006711975.2
XP_006712038.1
15
2386
XM_011532719.1
XP_011531021.1
16
2621
XM_011532720.2
XP_011531022.1
16
2630
XM_011532721.2
XP_011531023.1
15
2422
XM_011532723.1
XP_011531025.1
13
2052
XM_017003701.1
XP_016859190.1
14
2222
XM_017003702.1
XP_016859191.1
14
2262
XM_017003703.1
XP_016859192.1
15
2450
XM_017003704.1
XP_016859193.1
17
2768
XM_017003705.1
XP_016859194.1
17
2755
XM_017003706.1
XP_016859195.1
16
2547
XM_017003707.1
XP_016859196.1
13
1985
XM_017003708.1
XP_016859197.1
13
2218
XM_017003709.1
XP_016859198.1
14
2184
XM_017003710.1
XP_016859199.1
13
2034
XM_017003711.1
XP_016859200.1
13
1985
XM_017003712.1
XP_016859201.1
13
2053
XM_017003713.1
XP_016859202.1
14
2212
XM_017003714.1
XP_016859203.1
10
1831
XM_017003715.1
XP_016859204.1
12
1815
GenBank mRNA
AF272350.1
-
10
1615
AF315591.1
-
13
2202
AK093847.1
-
14
2303
AK304198.1
-
13
2112
BC024218.2
-
2
448
BC112046.1
-
13
2205
8327514
BC112048.1
-
13
2205
8327516
BC143550.1
-
12
1968
9052061
BC143552.1
-
13
2093
D87078.3
-
13
2179
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
posttranscriptional regulation of gene expression
stress granule assembly
regulation of mRNA stability
regulation of chromosome segregation
regulation of gene silencing by miRNA
positive regulation of RIG-I signaling pathway
positive regulation of gene silencing by miRNA
RNA binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-v5ndd:80/100.66.76.213:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline