Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015332.3 | NP_056147.2 | 5, 6 | 1161 | ||
XM_017011909.1 | XP_016867398.1 | 6, 7 | 1187 | |||
XM_017011910.1 | XP_016867399.1 | 6, 7 | 1045 | |||
GenBank mRNA | AB028991.1 | - | 5, 6 | 955 | ||
AK024226.1 | - | 4, 5 | 669 | |||
AK127163.1 | - | 562 | ||||
AK307613.1 | - | 3, 4 | 468 | |||
BC003691.2 | - | 5, 6 | 1022 | 3453305 | ||
BC011673.2 | - | 4, 5 | 781 | |||
BC017657.1 | - | 6, 7 | 1110 | |||
BC035014.2 | - | 5, 6 | 1059 | 4812229 | ||
EU350932.1 | - | 5, 6 | 979 |