Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015340.3 | NP_056155.1 | 13 | 1500 | ||
XM_005265006.1 | XP_005265063.1 | 12 | 1456 | |||
XM_011533554.2 | XP_011531856.1 | 13 | 1519 | |||
XM_017006042.1 | XP_016861531.1 | 13 | 1519 | |||
GenBank mRNA | AK025465.1 | - | 7 | 800 | ||
AK308146.1 | - | 11 | 1324 | |||
BC025989.2 | - | 13 | 1507 | 4822546 | ||
D21851.1 | - | 13 | 1500 |