Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5K1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195733.1 | NP_001182662.1 | 6 | 637 | ||
NM_001300849.1 | NP_001287778.1 | 6 | 650 | |||
NM_012398.2 | NP_036530.1 | 6 | 637 | |||
XM_011527845.1 | XP_011526147.1 | 6 | 644 | |||
XM_011527846.1 | XP_011526148.1 | 6 | 644 | |||
XM_011527848.1 | XP_011526150.1 | 6 | 644 | |||
XM_011527849.1 | XP_011526151.1 | 6 | 644 | |||
XM_011527850.2 | XP_011526152.1 | 6 | 603 | |||
XM_017026540.1 | XP_016882029.1 | 6 | 644 | |||
GenBank mRNA | AB011161.1 | - | 6 | 605 | ||
AK301309.1 | - | 6 | 620 | |||
AK315912.1 | - | 6 | 623 | |||
FJ965536.1 | - | 6 | 548 | |||
FJ965537.1 | - | 6 | 548 |