Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP13A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141973.2 | NP_001135445.1 | 16 | 1948 | ||
NM_001141974.2 | NP_001135446.1 | 16 | 1948 | |||
NM_022089.3 | NP_071372.1 | 16 | 1963 | |||
XM_005245810.1 | XP_005245867.1 | 16 | 1960 | |||
XM_005245811.1 | XP_005245868.1 | 16 | 1948 | |||
XM_005245812.1 | XP_005245869.1 | 16 | 1936 | |||
XM_005245815.1 | XP_005245872.1 | 16 | 1963 | |||
XM_006710512.1 | XP_006710575.1 | 16 | 1945 | |||
XM_006710513.1 | XP_006710576.1 | 16 | 1921 | |||
XM_011541128.1 | XP_011539430.1 | 16 | 1948 | |||
XM_017000844.1 | XP_016856333.1 | 16 | 1948 | |||
XM_017000845.1 | XP_016856334.1 | 16 | 1945 | |||
XM_017000846.1 | XP_016856335.1 | 16 | 1921 | |||
XM_017000847.1 | XP_016856336.1 | 16 | 1918 | |||
XM_017000848.1 | XP_016856337.1 | 16 | 1963 | |||
XM_017000849.1 | XP_016856338.1 | 16 | 1948 | |||
GenBank mRNA | AK075310.1 | - | 6 | 745 | ||
AK290210.1 | - | 16 | 1904 | |||
AL354615.1 | - | 16 | 1763 | |||
AY461712.1 | - | 16 | 1762 | |||
BC030267.1 | - | 16 | 1891 | 5240813 | ||
BC034575.1 | - | 13 | 1454 |