Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012240.2 | NP_036372.1 | 2 | 390 | ||
XM_006719308.2 | XP_006719371.1 | 2 | 1076 | |||
XM_006719309.3 | XP_006719372.1 | 2 | 572 | |||
GenBank mRNA | AF083109.1 | - | 2 | 351 | ||
BC109319.3 | - | 2 | 351 | 40036789 | ||
BC109320.3 | - | 2 | 351 | 40036792 | ||
BP232525.1 | - | 2 | 390 |