Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLYCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012213.2 | NP_036345.2 | 3 | 725 | ||
GenBank mRNA | AF090834.1 | - | 3 | 705 | ||
AF097832.2 | - | 3 | 643 | |||
AF153679.1 | - | 3 | 683 | |||
BC000286.1 | - | 3 | 677 | 3357140 | ||
BC052592.1 | - | 3 | 725 | 6421051 |