Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193640.1 | NP_001180569.1 | 4, 5 | 2007 | ||
NM_001257965.1 | NP_001244894.1 | 8, 9 | 2266 | |||
NM_201253.2 | NP_957705.1 | 6, 7 | 2343 | |||
NR_047563.1 | 7, 8 | 2135 | ||||
NR_047564.1 | 6, 7 | 2343 | ||||
XM_011509365.2 | XP_011507667.1 | 6, 7 | 2268 | |||
XM_011509367.1 | XP_011507669.1 | 6, 7 | 2268 | |||
XM_011509369.2 | XP_011507671.1 | 2, 3 | 1056 | |||
XM_017000851.1 | XP_016856340.1 | 5, 6 | 2035 | |||
XM_017000852.1 | XP_016856341.1 | 6, 7 | 2268 | |||
GenBank mRNA | AF154671.1 | - | 6, 7 | 2269 | ||
AJ748821.1 | - | 4, 5 | 1798 | |||
AK289733.1 | - | 6, 7 | 2288 | |||
AK299367.1 | - | 6, 7 | 1418 | |||
AK302818.1 | - | 2, 3 | 1130 | |||
AK302826.1 | - | 8, 9 | 2266 | |||
AK307844.1 | - | 3, 4 | 912 | |||
AY043324.1 | - | 6, 7 | 2269 | |||
AY043325.1 | - | 6, 7 | 2269 | |||
BC136271.1 | - | 6, 7 | 2182 | 9020258 | ||
BC143726.1 | - | 7, 8 | 1974 | |||
BX640729.1 | - | 2, 3 | 1136 |