Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193640.1 | NP_001180569.1 | 7 | 3130 | ||
NM_001257965.1 | NP_001244894.1 | 12 | 3524 | |||
NM_201253.2 | NP_957705.1 | 9 | 3466 | |||
NR_047563.1 | 10 | 3258 | ||||
NR_047564.1 | 9 | 3466 | ||||
XM_011509365.2 | XP_011507667.1 | 9 | 3391 | |||
XM_011509367.1 | XP_011507669.1 | 9 | 3391 | |||
XM_011509369.2 | XP_011507671.1 | 5 | 2179 | |||
XM_017000851.1 | XP_016856340.1 | 8 | 3158 | |||
XM_017000852.1 | XP_016856341.1 | 10 | 3526 | |||
GenBank mRNA | AF154671.1 | - | 9 | 3392 | ||
AJ748821.1 | - | 7 | 2921 | |||
AK289733.1 | - | 9 | 3411 | |||
AK302818.1 | - | 5 | 2253 | |||
AK302826.1 | - | 12 | 3524 | |||
AK307844.1 | - | 6 | 2035 | |||
AY043324.1 | - | 9 | 3392 | |||
AY043325.1 | - | 9 | 3392 | |||
BC136271.1 | - | 9 | 3305 | 9020258 | ||
BC143726.1 | - | 10 | 3097 | |||
BX640729.1 | - | 5 | 2259 |