Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286730.1 | NP_001273659.1 | 15 | 2155 | ||
NM_080546.4 | NP_536856.2 | 15 | 2155 | |||
XM_005251855.4 | XP_005251912.1 | 15 | 2160 | |||
XM_006717027.3 | XP_006717090.1 | 15 | 1968 | |||
XM_006717028.3 | XP_006717091.1 | 15 | 1968 | |||
XM_006717029.2 | XP_006717092.1 | 15 | 1968 | |||
XM_017014559.1 | XP_016870048.1 | 15 | 2143 | |||
XM_017014560.1 | XP_016870049.1 | 16 | 2133 | |||
GenBank mRNA | AJ272365.1 | - | 15 | 2108 | ||
AJ420812.1 | - | 15 | 1941 | |||
AK022549.1 | - | 11 | 1508 | |||
AK124926.1 | - | 10 | 2135 | |||
BC018213.1 | - | 4 | 470 | |||
BC049203.1 | - | 15 | 1908 | 5492167 |