Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243013.1 | NP_001229942.1 | 2 | 428 | ||
NM_001243014.1 | NP_001229943.1 | 2 | 428 | |||
NM_019624.3 | NP_062570.1 | 2 | 428 | |||
NM_019625.3 | NP_062571.1 | 2 | 428 | |||
NM_203444.3 | NP_982269.2 | 2 | 428 | |||
XM_005253558.2 | XP_005253615.1 | 2 | 382 | |||
XM_011538095.2 | XP_011536397.1 | 2 | 651 | |||
XM_011538096.2 | XP_011536398.1 | 2 | 447 | |||
XM_017019103.1 | XP_016874592.1 | 3 | 489 | |||
GenBank mRNA | AB040953.2 | - | 2 | 2957 | ||
AB045381.2 | - | 1 | 177 | |||
AB112582.1 | - | 2 | 379 | |||
AB112583.1 | - | 2 | 379 | |||
AB177852.1 | - | 2 | 340 | |||
AF216494.1 | - | 2 | 416 | |||
AK027624.1 | - | 2 | 321 | |||
AK304295.1 | - | 2 | 424 | |||
BC017348.2 | - | 1 | 143 | 5014247 | ||
BC064384.1 | - | 2 | 696 | 5744451 | ||
DA796102.1 | - | 2 | 428 |