Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080284.2 | NP_525023.2 | 32, 33 | 4360 | ||
XM_011524574.2 | XP_011522876.1 | 26, 27 | 3688 | |||
XM_017024404.1 | XP_016879893.1 | 32, 33 | 4672 | |||
XM_017024405.1 | XP_016879894.1 | 32, 33 | 4672 | |||
XM_017024406.1 | XP_016879895.1 | 28, 29 | 3640 | |||
XM_017024407.1 | XP_016879896.1 | 29, 30 | 3744 | |||
XM_017024408.1 | XP_016879897.1 | 18, 19 | 2233 | |||
GenBank mRNA | AF373290.1 | - | 31, 32 | 4188 | ||
AK097296.1 | - | 14, 15 | 1787 | |||
AK125487.1 | - | 11, 12 | 1452 | |||
AY028898.1 | - | 32, 33 | 4360 | |||
BC125231.1 | - | 32, 33 | 4359 | |||
BC125232.1 | - | 32, 33 | 4359 | |||
U66680.1 | - | 3, 4 | 377 |