Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPTXR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014293.3 | NP_055108.2 | 3 | 1196 | ||
GenBank mRNA | AL161974.1 | - | 3 | 510 |