Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301140.1 | NP_001288069.1 | 3, 4 | 1509 | ||
NM_001301141.1 | NP_001288070.1 | 3, 4 | 1509 | |||
NM_001320042.1 | NP_001306971.1 | 3, 4 | 1509 | |||
NM_014301.4 | NP_055116.1 | 4, 5 | 1605 | |||
NM_213595.3 | NP_998760.1 | 3, 4 | 1509 | |||
NR_135127.1 | 4, 5 | 1605 | ||||
XM_011538100.2 | XP_011536402.1 | 4, 5 | 1605 | |||
GenBank mRNA | AK000574.1 | - | 3, 4 | 352 | ||
AK022758.1 | - | 2, 3 | 1933 | |||
AK057251.1 | - | 3, 4 | 1509 | |||
AK293202.1 | - | 3, 4 | 362 | |||
AK297862.1 | - | 3, 4 | 381 | |||
AY009127.1 | - | 4, 5 | 462 | |||
AY009128.1 | - | 3, 4 | 366 | |||
BC011906.2 | - | 3, 4 | 365 | 4136262 | ||
BC061903.1 | - | 3, 4 | 358 | 4183674 | ||
BG702203.1 | - | 3, 4 | 360 | |||
BI597126.1 | - | 3, 4 | 460 | |||
BM423850.1 | - | 3, 4 | 355 | |||
BM921073.1 | - | 3, 4 | 450 | |||
BP378051.1 | - | 3, 4 | 381 | |||
BQ020993.1 | - | 2 | 637 | |||
BU602914.1 | - | 3, 4 | 370 | |||
DB095495.1 | - | 3, 4 | 376 | |||
DB493673.1 | - | 3, 4 | 439 | |||
EU329002.1 | - | 4, 5 | 462 | |||
U47101.1 | - | 2, 3 | 212 |