Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015346.3 | NP_056161.2 | 6 | 1099 | ||
XM_011536609.2 | XP_011534911.1 | 6 | 1100 | |||
XM_017021124.1 | XP_016876613.1 | 6 | 1100 | |||
XM_017021125.1 | XP_016876614.1 | 6 | 1100 | |||
XM_017021126.1 | XP_016876615.1 | 6 | 1803 | |||
GenBank mRNA | AB425197.1 | - | 6 | 1087 | ||
AK304428.1 | - | 6 | 1095 | |||
AK308233.1 | - | 6 | 1099 | |||
BC144594.1 | - | 6 | 1090 | |||
BX537886.1 | - | 6 | 1093 | |||
BX538025.1 | - | 2 | 592 | |||
BX648036.1 | - | 6 | 1096 | |||
BX648683.1 | - | 6 | 1097 | |||
CR749276.1 | - | 7 | 1034 | |||
CR749365.1 | - | 6 | 1098 |