Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUZ12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321207.1 | NP_001308136.1 | 11 | 1548 | ||
NM_015355.3 | NP_056170.2 | 12 | 1617 | |||
XM_006721794.3 | XP_006721857.1 | 10 | 1059 | |||
XM_017024409.1 | XP_016879898.1 | 11 | 1273 | |||
GenBank mRNA | AK074333.1 | - | 7 | 854 | ||
AK290014.1 | - | 12 | 1588 | |||
BC015704.1 | - | 12 | 1459 | 3908024 | ||
BC018583.1 | - | 11 | 1491 | |||
D63881.2 | - | 12 | 1574 |