Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128431.2 | NP_001121903.1 | 2 | 314 | ||
NM_001135153.1 | NP_001128625.1 | 2 | 355 | |||
NM_001135154.1 | NP_001128626.1 | 2 | 314 | |||
NM_015359.4 | NP_056174.2 | 2 | 314 | |||
XM_005273465.2 | XP_005273522.1 | 3 | 402 | |||
XM_005273466.4 | XP_005273523.1 | 2 | 386 | |||
XM_006716323.2 | XP_006716386.1 | 4 | 600 | |||
XM_006716324.2 | XP_006716387.1 | 3 | 391 | |||
XM_011544478.2 | XP_011542780.1 | 3 | 466 | |||
XM_017013292.1 | XP_016868781.1 | 3 | 482 | |||
XM_017013293.1 | XP_016868782.1 | 4 | 600 | |||
XM_017013294.1 | XP_016868783.1 | 4 | 509 | |||
XM_017013295.1 | XP_016868784.1 | 2 | 519 | |||
GenBank mRNA | AK172810.1 | - | 2 | 285 | ||
AK295807.1 | - | 2 | 230 | |||
BC015770.1 | - | 2 | 276 | 4865469 | ||
D31887.1 | - | 2 | 258 | |||
DC323208.1 | - | 2 | 314 | |||
DC423882.1 | - | 2 | 356 | |||
KU178568.1 | - | 1 | 139 | |||
KU178569.1 | - | 1 | 139 | |||
KU178570.1 | - | 1 | 139 | |||
KU178571.1 | - | 1 | 139 |