Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199281.1 | NP_001186210.1 | 15, 16 | 2475 | ||
NM_001201429.1 | NP_001188358.1 | 14, 15 | 2019 | |||
NM_012295.3 | NP_036427.1 | 15, 16 | 2169 | |||
XM_005261415.2 | XP_005261472.1 | 14, 15 | 2116 | |||
XM_005261416.2 | XP_005261473.1 | 14, 15 | 2116 | |||
XM_005261417.2 | XP_005261474.1 | 14, 15 | 2116 | |||
XM_005261419.2 | XP_005261476.1 | 14, 15 | 2157 | |||
XM_006724179.1 | XP_006724242.1 | 14, 15 | 2116 | |||
XM_006724180.1 | XP_006724243.1 | 14, 15 | 2116 | |||
XM_006724181.1 | XP_006724244.1 | 14, 15 | 2116 | |||
XM_011530029.2 | XP_011528331.1 | 14, 15 | 2116 | |||
XM_011530030.2 | XP_011528332.1 | 14, 15 | 2116 | |||
XM_017028680.1 | XP_016884169.1 | 14, 15 | 2116 | |||
XM_017028681.1 | XP_016884170.1 | 13, 14 | 1966 | |||
XM_017028682.1 | XP_016884171.1 | 14, 15 | 2037 | |||
XM_017028683.1 | XP_016884172.1 | 13, 14 | 1966 | |||
GenBank mRNA | AB002328.2 | - | 15, 16 | 2392 | ||
AF072441.1 | - | 15, 16 | 2170 | |||
BC054497.1 | - | 14, 15 | 1986 | 4521929 |