Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166212.1 | NP_001159684.1 | 3 | 326 | ||
NM_013246.2 | NP_037378.1 | 3 | 407 | |||
GenBank mRNA | AF172854.1 | - | 3 | 256 | ||
AF176911.1 | - | 3 | 300 | |||
AK298052.1 | - | 3 | 326 | |||
AY049779.1 | - | 3 | 213 | |||
BC012939.1 | - | 3 | 283 | 4453813 | ||
BC066229.1 | - | 3 | 215 | 7002179 | ||
BC066230.1 | - | 3 | 215 | 7002180 | ||
BC066231.1 | - | 3 | 215 | 7002181 | ||
BM846622.1 | - | 3 | 409 | |||
DC393345.1 | - | 3 | 326 |