Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012329.2 | NP_036461.2 | 2, 3 | 399 | ||
XM_006721795.2 | XP_006721858.1 | 2, 3 | 412 | |||
GenBank mRNA | AK312760.1 | - | 2, 3 | 278 | ||
AY424289.1 | - | 2, 3 | 379 | |||
BC026324.1 | - | 2, 3 | 371 | 4794375 | ||
BI546449.1 | - | 2, 3 | 411 | |||
CR541893.1 | - | 2, 3 | 114 | |||
X85750.1 | - | 2, 3 | 195 |