Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142633.2 | NP_001136105.1 | 12 | 1944 | ||
NM_001251851.1 | NP_001238780.1 | 12 | 2006 | |||
NM_001251852.1 | NP_001238781.1 | 11 | 1828 | |||
NM_001251853.1 | NP_001238782.1 | 11 | 1790 | |||
NM_001251855.1 | NP_001238784.1 | 11 | 1769 | |||
NM_014308.3 | NP_055123.2 | 12 | 1881 | |||
XM_005256580.3 | XP_005256637.1 | 12 | 1939 | |||
GenBank mRNA | AB028925.1 | - | 11 | 1814 | ||
AF128881.1 | - | 12 | 1881 | |||
AK314186.1 | - | 12 | 1939 | |||
BC028212.1 | - | 11 | 1760 | 5215094 |