Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012269.2 | NP_036401.2 | 3 | 1398 | ||
XM_011515990.2 | XP_011514292.1 | 3 | 1474 | |||
XM_011515991.2 | XP_011514293.1 | 3 | 1056 | |||
XM_017011911.1 | XP_016867400.1 | 4 | 1101 | |||
GenBank mRNA | AB470346.1 | - | 2 | 830 | ||
AF009010.1 | - | 3 | 1401 | |||
BC104788.1 | - | 2 | 836 | 8143791 | ||
BC104790.1 | - | 2 | 836 | 8143793 |