Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303007.1 | NP_001289936.1 | 5 | 1711 | ||
NM_001303008.1 | NP_001289937.1 | 6 | 811 | |||
NM_013974.2 | NP_039268.1 | 6 | 886 | |||
XM_011514448.2 | XP_011512750.1 | 6 | 962 | |||
GenBank mRNA | AF070667.1 | - | 6 | 883 | ||
AF087894.1 | - | 6 | 883 | |||
AK026191.1 | - | 5 | 1238 | |||
AK098671.1 | - | 6 | 873 | |||
BC001435.2 | - | 6 | 798 | 3138832 | ||
CB242623.1 | - | 2 | 464 | |||
EB387244.1 | - | 6 | 785 | |||
FJ224290.1 | - | 6 | 883 |