Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308213.1 | NP_001295142.1 | 7 | 906 | ||
NM_001308214.1 | NP_001295143.1 | 5 | 735 | |||
NM_001308215.1 | NP_001295144.1 | 5 | 645 | |||
NM_001308218.1 | NP_001295147.1 | 3 | 537 | |||
NM_001308221.1 | NP_001295150.1 | 2 | 303 | |||
NM_001308223.1 | NP_001295152.1 | 2 | 303 | |||
NM_012279.3 | NP_036411.1 | 6 | 831 | |||
NR_131773.1 | 5 | 686 | ||||
NR_131774.1 | 4 | 500 | ||||
XM_005265860.2 | XP_005265917.1 | 7 | 899 | |||
XM_005265864.3 | XP_005265921.1 | 6 | 665 | |||
XM_011534493.1 | XP_011532795.1 | 7 | 899 | |||
XM_017009294.1 | XP_016864783.1 | 6 | 879 | |||
XM_017009295.1 | XP_016864784.1 | 6 | 831 | |||
XM_017009296.1 | XP_016864785.1 | 6 | 831 | |||
XM_017009300.1 | XP_016864789.1 | 6 | 1169 | |||
XM_017009301.1 | XP_016864790.1 | 5 | 645 | |||
XM_017009303.1 | XP_016864792.1 | 4 | 942 | |||
GenBank mRNA | AF083340.1 | - | 6 | 804 | ||
AK026627.1 | - | 6 | 799 | |||
AK131469.1 | - | 7 | 851 | |||
AK295549.1 | - | 5 | 711 | |||
AK297448.1 | - | 3 | 521 | |||
AK297584.1 | - | 2 | 287 | |||
AK300065.1 | - | 5 | 650 | |||
AK300738.1 | - | 4 | 445 | |||
AL359576.1 | - | 6 | 779 | |||
BC007775.2 | - | 6 | 786 | 4301820 | ||
BU860103.1 | - | 5 | 620 | |||
CR749692.1 | - | 6 | 779 | |||
DR003225.1 | - | 2 | 306 |