Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNDBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012142.4 | NP_036274.3 | 4 | 517 | ||
NR_027513.2 | 3 | 437 | ||||
NR_027514.2 | 4 | 517 | ||||
NR_045998.1 | 3 | 437 | ||||
NR_045999.1 | 2 | 377 | ||||
XM_006720448.1 | XP_006720511.1 | 2 | 439 | |||
GenBank mRNA | AF082569.1 | - | 4 | 376 | ||
AF087852.1 | - | 4 | 362 | |||
AF113535.1 | - | 2 | 326 | |||
AF132034.1 | - | 4 | 332 | |||
AF246144.1 | - | 4 | 334 | |||
AK075146.1 | - | 4 | 346 | |||
AK075296.1 | - | 2 | 331 | |||
AK128849.1 | - | 1 | 787 | |||
AK225143.1 | - | 4 | 332 | |||
AK225645.1 | - | 4 | 346 | |||
AK290665.1 | - | 4 | 346 | |||
AK295017.1 | - | 4 | 346 | |||
AK308817.1 | - | 3 | 941 | |||
AL552334.3 | - | 4 | 456 | |||
BC009689.1 | - | 4 | 360 | 3896421 | ||
BI911845.1 | - | 288 | ||||
BQ439439.1 | - | 2 | 191 | |||
CR450331.1 | - | 4 | 286 | |||
DA506028.1 | - | 4 | 506 |