Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014313.3 | NP_055128.1 | 3 | 292 | ||
XM_005245817.1 | XP_005245874.1 | 3 | 256 | |||
XM_011541159.1 | XP_011539461.1 | 3 | 486 | |||
GenBank mRNA | AF081282.1 | - | 3 | 219 | ||
AF087880.1 | - | 3 | 280 | |||
AK299180.1 | - | 3 | 256 | |||
AK312588.1 | - | 3 | 256 | |||
AL136627.1 | - | 3 | 274 | |||
AY071927.1 | - | 3 | 269 | |||
AY358650.1 | - | 3 | 255 | |||
BC007341.2 | - | 3 | 240 | 3688672 | ||
BC066908.1 | - | 3 | 231 | 4817784 | ||
JX644905.1 | - | 3 | 292 | |||
JX644906.1 | - | 3 | 292 |