Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014314.3 | NP_055129.2 | 13 | 1973 | ||
GenBank mRNA | AF038963.1 | - | 13 | 1972 | ||
AK125989.1 | - | 14 | 2178 | |||
AK301678.1 | - | 12 | 1696 | |||
AK315040.1 | - | 13 | 1844 | |||
AL137608.1 | - | 3 | 234 | |||
BC132786.1 | - | 13 | 1921 | 40146808 | ||
BC136610.1 | - | 13 | 1921 | 9020600 | ||
BX647917.1 | - | 12 | 1710 |