Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326380.1 | NP_001313309.1 | 2 | 230 | ||
NM_001326381.1 | NP_001313310.1 | 2 | 461 | |||
NM_014320.2 | NP_055135.1 | 2 | 461 | |||
XM_017010631.1 | XP_016866120.1 | 2 | 434 | |||
GenBank mRNA | AF117616.1 | - | 2 | 439 | ||
AF411610.1 | - | 2 | 101 | |||
AK057928.1 | - | 2 | 376 | |||
AY427823.1 | - | 2 | 343 | |||
BC008205.1 | - | 2 | 325 | 3869026 | ||
BC010290.1 | - | 2 | 338 | 3445763 | ||
BC016806.1 | - | 2 | 441 | |||
BC016807.1 | - | 2 | 441 | |||
BC093037.1 | - | 2 | 324 | 30333215 | ||
BF974311.1 | - | 461 |