Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167595.1 | NP_001161067.1 | 1 | 162 | ||
NM_014324.5 | NP_055139.4 | 1 | 162 | |||
NM_203382.2 | NP_976316.1 | 1 | 162 | |||
GenBank mRNA | AF047020.1 | - | 1 | 154 | ||
AF158378.1 | - | 1 | 131 | |||
AJ130733.1 | - | 1 | 154 | |||
AK000912.1 | - | 1 | 147 | |||
AK022765.1 | - | 1 | 132 | |||
AK222831.1 | - | 1 | 134 | |||
AK225749.1 | - | 1 | 171 | |||
AK292988.1 | - | 1 | 112 | |||
AL545355.3 | - | 1 | 75 | |||
AY330488.1 | - | 1 | 75 | |||
AY330489.1 | - | 1 | 75 | |||
AY935981.1 | - | 1 | 75 | |||
BC009471.1 | - | 1 | 78 | 2958112 | ||
BG708726.1 | - | 1 | 98 | |||
BI550368.1 | - | 1 | 200 | |||
BT007193.1 | - | 1 | 66 | |||
DA156454.1 | - | 1 | 165 | |||
EF560721.1 | - | 1 | 85 | |||
FJ498906.1 | - | 1 | 78 | |||
FJ498907.1 | - | 1 | 78 | |||
FJ498908.1 | - | 1 | 78 |