Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014326.3 | NP_055141.2 | 8 | 820 | ||
XM_011521413.2 | XP_011519715.1 | 7 | 795 | |||
XM_011521414.1 | XP_011519716.1 | 7 | 795 | |||
XM_011521415.2 | XP_011519717.1 | 7 | 795 | |||
XM_011521416.2 | XP_011519718.1 | 8 | 968 | |||
XM_011521417.1 | XP_011519719.1 | 7 | 795 | |||
XM_011521421.2 | XP_011519723.1 | 6 | 605 | |||
XM_017022043.1 | XP_016877532.1 | 6 | 564 | |||
XM_017022044.1 | XP_016877533.1 | 7 | 795 | |||
XM_017022045.1 | XP_016877534.1 | 7 | 795 | |||
XM_017022046.1 | XP_016877535.1 | 7 | 795 | |||
XM_017022047.1 | XP_016877536.1 | 6 | 605 | |||
XM_017022048.1 | XP_016877537.1 | 5 | 554 | |||
XM_017022049.1 | XP_016877538.1 | 6 | 601 | |||
XM_017022050.1 | XP_016877539.1 | 5 | 406 | |||
XM_017022051.1 | XP_016877540.1 | 5 | 552 | |||
XM_017022052.1 | XP_016877541.1 | 2 | 1088 | |||
GenBank mRNA | AB018001.1 | - | 8 | 819 | ||
AF052941.1 | - | 8 | 820 | |||
BC114506.1 | - | 7 | 809 | 40005390 | ||
BC114951.1 | - | 7 | 717 | 40005391 | ||
GU056176.1 | - | 8 | 820 |