Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
UniGene ID:
Hs.446240
Species:
Human
siRNA ID
HSS119059
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281771.2
NP_001268700.1
11
1247
NM_001281772.2
NP_001268701.1
13
1446
NM_001281773.2
NP_001268702.1
12
1393
NM_001281774.2
NP_001268703.1
12
1393
NM_001281775.2
NP_001268704.1
11
1322
NM_001281776.2
NP_001268705.1
11
1322
NM_001281777.2
NP_001268706.1
11
1247
NM_001281778.2
NP_001268707.1
11
1247
NM_001281779.2
NP_001268708.1
12
1453
NM_001281780.2
NP_001268709.1
12
1453
NM_001281781.2
NP_001268710.1
11
1247
NM_001281782.2
NP_001268711.1
11
1247
NM_001281783.2
NP_001268712.1
11
1322
NM_001281784.2
NP_001268713.1
11
1247
NM_012408.5
NP_036540.3
11
1322
NM_183047.3
NP_898868.1
11
1322
NM_183048.3
NP_898869.1
11
1247
XM_005260356.4
XP_005260413.1
11
1359
XM_005260358.2
XP_005260415.1
10
1101
XM_005260360.4
XP_005260417.1
11
1824
XM_005260362.2
XP_005260419.1
11
1099
XM_005260364.2
XP_005260421.1
11
1084
XM_005260366.2
XP_005260423.1
11
1115
XM_006723760.3
XP_006723823.1
11
1497
XM_006723762.3
XP_006723825.1
11
1198
XM_006723763.1
XP_006723826.1
11
1113
XM_006723766.1
XP_006723829.1
11
1057
XM_011528749.2
XP_011527051.1
11
2003
XM_011528751.1
XP_011527053.1
11
1107
XM_011528752.1
XP_011527054.1
10
1089
XM_011528753.1
XP_011527055.1
12
1118
XM_011528754.1
XP_011527056.1
11
1045
XM_017027760.1
XP_016883249.1
10
1104
XM_017027761.1
XP_016883250.1
11
1570
XM_017027762.1
XP_016883251.1
13
2337
XM_017027763.1
XP_016883252.1
11
1250
XM_017027764.1
XP_016883253.1
11
1769
XM_017027765.1
XP_016883254.1
11
1099
XM_017027766.1
XP_016883255.1
11
1099
XM_017027767.1
XP_016883256.1
11
1914
XM_017027768.1
XP_016883257.1
11
1174
XM_017027769.1
XP_016883258.1
11
1084
XM_017027770.1
XP_016883259.1
12
1118
XM_017027771.1
XP_016883260.1
11
1084
XM_017027772.1
XP_016883261.1
11
1057
XM_017027773.1
XP_016883262.1
11
1099
XM_017027774.1
XP_016883263.1
10
1077
XM_017027775.1
XP_016883264.1
11
1045
XM_017027776.1
XP_016883265.1
11
1084
XM_017027777.1
XP_016883266.1
10
1077
XM_017027778.1
XP_016883267.1
10
1089
XM_017027779.1
XP_016883268.1
10
1089
XM_017027780.1
XP_016883269.1
10
1104
GenBank mRNA
AB032951.1
-
11
1297
AF454056.1
-
11
1139
AK122966.1
-
10
943
AK225857.1
-
11
1130
AK294511.1
-
13
1256
AK295747.1
-
11
1206
AK299899.1
-
12
1205
AK309228.1
-
11
1116
AL137703.1
-
10
1059
BC030721.1
-
8
822
BC136608.1
-
11
1078
9020598
BC144289.1
-
11
1078
9052809
BC146802.1
-
11
1088
8860316
BX641005.1
-
12
1338
DQ082998.1
-
10
1103
DQ082999.1
-
10
1103
DQ368669.1
-
10
1103
DQ368670.1
-
10
1103
DQ368671.1
-
10
1028
DQ368672.1
-
10
1028
DQ368673.1
-
10
1103
DQ368674.1
-
10
1028
DQ368675.1
-
11
1234
DQ368676.1
-
11
1234
DQ368677.1
-
10
1028
DQ368678.1
-
10
1028
DQ368679.1
-
10
1103
DQ368680.1
-
10
1028
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription corepressor activity
chromatin binding
transcription cofactor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline