Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
UniGene ID:
Hs.446240
Species:
Human
siRNA ID
HSS119060
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281771.2
NP_001268700.1
13
1902
NM_001281772.2
NP_001268701.1
15
2101
NM_001281773.2
NP_001268702.1
14
2048
NM_001281774.2
NP_001268703.1
14
2048
NM_001281775.2
NP_001268704.1
13
1977
NM_001281776.2
NP_001268705.1
13
1977
NM_001281777.2
NP_001268706.1
13
1902
NM_001281778.2
NP_001268707.1
13
1902
NM_001281779.2
NP_001268708.1
14
2108
NM_001281780.2
NP_001268709.1
14
2108
NM_001281781.2
NP_001268710.1
12
1761
NM_001281782.2
NP_001268711.1
13
1902
NM_001281783.2
NP_001268712.1
13
1977
NM_001281784.2
NP_001268713.1
12
1761
NM_012408.5
NP_036540.3
13
1977
NM_183047.3
NP_898868.1
13
1977
NM_183048.3
NP_898869.1
13
1902
XM_005260356.4
XP_005260413.1
13
2014
XM_005260358.2
XP_005260415.1
12
1756
XM_005260360.4
XP_005260417.1
13
2479
XM_005260362.2
XP_005260419.1
13
1754
XM_005260364.2
XP_005260421.1
13
1739
XM_005260366.2
XP_005260423.1
13
1770
XM_006723760.3
XP_006723823.1
13
2152
XM_006723762.3
XP_006723825.1
13
1853
XM_006723763.1
XP_006723826.1
13
1768
XM_006723766.1
XP_006723829.1
13
1712
XM_011528749.2
XP_011527051.1
13
2658
XM_011528751.1
XP_011527053.1
13
1762
XM_011528752.1
XP_011527054.1
11
1603
XM_011528753.1
XP_011527055.1
14
1773
XM_011528754.1
XP_011527056.1
13
1700
XM_017027760.1
XP_016883249.1
12
1759
XM_017027761.1
XP_016883250.1
13
2225
XM_017027762.1
XP_016883251.1
15
2992
XM_017027763.1
XP_016883252.1
13
1905
XM_017027764.1
XP_016883253.1
13
2424
XM_017027765.1
XP_016883254.1
13
1754
XM_017027766.1
XP_016883255.1
13
1754
XM_017027767.1
XP_016883256.1
13
2569
XM_017027768.1
XP_016883257.1
13
1829
XM_017027769.1
XP_016883258.1
13
1739
XM_017027770.1
XP_016883259.1
14
1773
XM_017027771.1
XP_016883260.1
13
1739
XM_017027772.1
XP_016883261.1
13
1712
XM_017027773.1
XP_016883262.1
13
1754
XM_017027774.1
XP_016883263.1
12
1732
XM_017027775.1
XP_016883264.1
13
1700
XM_017027776.1
XP_016883265.1
12
1598
XM_017027777.1
XP_016883266.1
12
1732
XM_017027778.1
XP_016883267.1
12
1744
XM_017027779.1
XP_016883268.1
12
1744
XM_017027780.1
XP_016883269.1
12
1759
GenBank mRNA
AB032951.1
-
13
1952
AF233453.1
-
2
178
AF273045.1
-
3
575
AF454056.1
-
13
1794
AK000275.1
-
13
1719
AK026151.1
-
3
522
AK074262.1
-
3
512
AK122966.1
-
12
1598
AK225857.1
-
13
1785
AK294511.1
-
15
1911
AK295747.1
-
13
1861
AK299899.1
-
14
1860
AK309228.1
-
13
1771
AL137703.1
-
12
1713
BC030721.1
-
10
1477
BC136608.1
-
13
1733
9020598
BC144289.1
-
13
1733
9052809
BC146802.1
-
13
1743
8860316
BX641005.1
-
14
1993
DQ082998.1
-
12
1758
DQ082999.1
-
12
1758
DQ368669.1
-
12
1758
DQ368670.1
-
12
1758
DQ368671.1
-
12
1683
DQ368672.1
-
12
1683
DQ368673.1
-
12
1758
DQ368674.1
-
12
1683
DQ368675.1
-
13
1889
DQ368676.1
-
13
1889
DQ368677.1
-
11
1542
DQ368678.1
-
12
1683
DQ368679.1
-
12
1758
DQ368680.1
-
11
1542
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription corepressor activity
chromatin binding
transcription cofactor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0