Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
UniGene ID:
Hs.446240
Species:
Human
siRNA ID
HSS119061
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281771.2
NP_001268700.1
8
919
NM_001281772.2
NP_001268701.1
10
1118
NM_001281773.2
NP_001268702.1
9
1065
NM_001281774.2
NP_001268703.1
9
1065
NM_001281775.2
NP_001268704.1
8
994
NM_001281776.2
NP_001268705.1
8
994
NM_001281777.2
NP_001268706.1
8
919
NM_001281778.2
NP_001268707.1
8
919
NM_001281779.2
NP_001268708.1
8
994
NM_001281780.2
NP_001268709.1
8
994
NM_001281781.2
NP_001268710.1
8
919
NM_001281782.2
NP_001268711.1
8
919
NM_001281783.2
NP_001268712.1
8
994
NM_001281784.2
NP_001268713.1
8
919
NM_012408.5
NP_036540.3
8
994
NM_183047.3
NP_898868.1
8
994
NM_183048.3
NP_898869.1
8
919
XM_005260356.4
XP_005260413.1
8
1031
XM_005260358.2
XP_005260415.1
7
773
XM_005260360.4
XP_005260417.1
8
1496
XM_005260362.2
XP_005260419.1
8
771
XM_005260364.2
XP_005260421.1
8
756
XM_005260366.2
XP_005260423.1
8
787
XM_006723760.3
XP_006723823.1
8
1169
XM_006723762.3
XP_006723825.1
8
870
XM_006723763.1
XP_006723826.1
8
785
XM_006723766.1
XP_006723829.1
8
729
XM_011528749.2
XP_011527051.1
8
1675
XM_011528751.1
XP_011527053.1
8
779
XM_011528752.1
XP_011527054.1
7
761
XM_011528753.1
XP_011527055.1
9
790
XM_011528754.1
XP_011527056.1
8
717
XM_017027760.1
XP_016883249.1
7
776
XM_017027761.1
XP_016883250.1
8
1242
XM_017027762.1
XP_016883251.1
10
2009
XM_017027763.1
XP_016883252.1
8
922
XM_017027764.1
XP_016883253.1
8
1441
XM_017027765.1
XP_016883254.1
8
771
XM_017027766.1
XP_016883255.1
8
771
XM_017027767.1
XP_016883256.1
8
1586
XM_017027768.1
XP_016883257.1
8
846
XM_017027769.1
XP_016883258.1
8
756
XM_017027770.1
XP_016883259.1
9
790
XM_017027771.1
XP_016883260.1
8
756
XM_017027772.1
XP_016883261.1
8
729
XM_017027773.1
XP_016883262.1
8
771
XM_017027774.1
XP_016883263.1
7
749
XM_017027775.1
XP_016883264.1
8
717
XM_017027776.1
XP_016883265.1
8
756
XM_017027777.1
XP_016883266.1
7
749
XM_017027778.1
XP_016883267.1
7
761
XM_017027779.1
XP_016883268.1
7
761
XM_017027780.1
XP_016883269.1
7
776
GenBank mRNA
AB032951.1
-
8
969
AF454056.1
-
8
811
AK000275.1
-
8
736
AK122966.1
-
7
615
AK225857.1
-
8
802
AK294511.1
-
10
928
AK295747.1
-
8
878
AK299899.1
-
9
877
AK309228.1
-
8
788
AL137703.1
-
7
731
BC030721.1
-
5
494
BC136608.1
-
8
750
9020598
BC144289.1
-
8
750
9052809
BC146802.1
-
8
760
8860316
BX641005.1
-
9
1010
DQ082998.1
-
7
775
DQ082999.1
-
7
775
DQ368669.1
-
7
775
DQ368670.1
-
7
775
DQ368671.1
-
7
700
DQ368672.1
-
7
700
DQ368673.1
-
7
775
DQ368674.1
-
7
700
DQ368675.1
-
7
775
DQ368676.1
-
7
775
DQ368677.1
-
7
700
DQ368678.1
-
7
700
DQ368679.1
-
7
775
DQ368680.1
-
7
700
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription corepressor activity
chromatin binding
transcription cofactor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline