Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012104.4 | NP_036236.1 | 1 | 644 | ||
NM_138971.3 | NP_620427.1 | 1 | 644 | |||
NM_138972.3 | NP_620428.1 | 1 | 644 | |||
NM_138973.3 | NP_620429.1 | 1 | 644 | |||
GenBank mRNA | AB032975.2 | - | 1 | 626 | ||
AB050436.1 | - | 1 | 186 | |||
AB050437.1 | - | 1 | 186 | |||
AB050438.1 | - | 1 | 186 | |||
AB089958.1 | - | 1 | 183 | |||
AF190725.1 | - | 1 | 636 | |||
AF200193.1 | - | 1 | 144 | |||
AF200343.1 | - | 1 | 183 | |||
AF201468.1 | - | 1 | 637 | |||
AF204943.1 | - | 1 | 183 | |||
AF338816.1 | - | 1 | 183 | |||
AF338817.1 | - | 1 | 183 | |||
BC036084.1 | - | 1 | 639 | 5311572 | ||
BC065492.1 | - | 1 | 644 | 6538154 |