Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012316.4 | NP_036448.1 | 8, 9 | 831 | ||
XM_005270711.3 | XP_005270768.1 | 8, 9 | 1192 | |||
GenBank mRNA | AF060543.1 | - | 8, 9 | 798 | ||
AK002111.1 | - | 8, 9 | 818 | |||
AK128609.1 | - | 8, 9 | 794 | |||
AK295615.1 | - | 8, 9 | 946 | |||
AK301714.1 | - | 8, 9 | 785 | |||
AK315490.1 | - | 8, 9 | 818 | |||
BC020520.1 | - | 8, 9 | 806 | 3913864 | ||
BT009843.1 | - | 7, 8 | 743 |