Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256732.1 | NP_001243661.1 | 13 | 1071 | ||
NM_001256733.1 | NP_001243662.1 | 12 | 987 | |||
NM_001256734.1 | NP_001243663.1 | 12 | 981 | |||
NM_001256735.1 | NP_001243664.1 | 12 | 957 | |||
NM_001256736.1 | NP_001243665.1 | 12 | 957 | |||
NM_012446.3 | NP_036578.2 | 13 | 1047 | |||
XM_011543297.2 | XP_011541599.1 | 13 | 986 | |||
XM_017009304.1 | XP_016864793.1 | 13 | 889 | |||
XM_017009305.1 | XP_016864794.1 | 14 | 978 | |||
XM_017009306.1 | XP_016864795.1 | 13 | 962 | |||
XM_017009307.1 | XP_016864796.1 | 12 | 775 | |||
XM_017009308.1 | XP_016864797.1 | 14 | 1150 | |||
XM_017009309.1 | XP_016864798.1 | 13 | 1214 | |||
XM_017009310.1 | XP_016864799.1 | 13 | 1362 | |||
GenBank mRNA | AF077048.1 | - | 13 | 911 | ||
AF161465.1 | - | 13 | 877 | |||
AK074537.1 | - | 13 | 842 | |||
AK293544.1 | - | 12 | 782 | |||
AK294864.1 | - | 12 | 752 | |||
AK297494.1 | - | 13 | 755 | |||
AK299858.1 | - | 12 | 825 | |||
AK312337.1 | - | 13 | 891 | |||
AL080076.1 | - | 13 | 895 | |||
BC017020.1 | - | 13 | 895 | 3913337 | ||
CR936787.1 | - | 12 | 802 | |||
DA202987.1 | - | 5 | 237 |