Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195797.1 | NP_001182726.1 | 2 | 301 | ||
NM_015364.4 | NP_056179.3 | 3 | 391 | |||
XM_011517508.2 | XP_011515810.1 | 3 | 324 | |||
XM_017013299.1 | XP_016868788.1 | 3 | 324 | |||
XM_017013300.1 | XP_016868789.1 | 2 | 355 | |||
GenBank mRNA | AB018549.1 | - | 3 | 402 | ||
AB446498.1 | - | 3 | 277 | |||
AF168121.1 | - | 3 | 298 | |||
BC020690.1 | - | 3 | 301 | 4767246 | ||
BG435728.1 | - | 3 | 392 |