Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014329.4 | NP_055144.3 | 23 | 3365 | ||
GenBank mRNA | AK128582.1 | - | 22 | 3839 | ||
AK291049.1 | - | 23 | 3133 | |||
BC043616.1 | - | 23 | 3268 | 6065418 | ||
BC053598.1 | - | 18 | 2505 | |||
BC064567.1 | - | 23 | 3262 | 5575195 | ||
L26339.1 | - | 23 | 3262 | |||
U17474.1 | - | 13 | 1902 |