Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242854.1 | NP_001229783.1 | 8 | 1278 | ||
NM_001242855.1 | NP_001229784.1 | 7 | 1038 | |||
NM_001242856.1 | NP_001229785.1 | 6 | 919 | |||
NM_012402.3 | NP_036534.1 | 8 | 1139 | |||
XM_005252840.4 | XP_005252897.1 | 8 | 1043 | |||
GenBank mRNA | AK026390.1 | - | 8 | 971 | ||
AK300855.1 | - | 8 | 1139 | |||
AK301972.1 | - | 8 | 1080 | |||
AK304512.1 | - | 6 | 786 | |||
AK310267.1 | - | 3 | 968 | |||
BC000392.2 | - | 8 | 961 | 2821666 | ||
U52522.1 | - | 8 | 955 | |||
X97567.1 | - | 6 | 774 |