Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012101.3 | NP_036233.2 | 6 | 1615 | ||
XM_005271490.1 | XP_005271547.1 | 6 | 792 | |||
XM_011542729.1 | XP_011541031.1 | 6 | 1639 | |||
XM_011542732.1 | XP_011541034.1 | 6 | 1639 | |||
XM_011542733.1 | XP_011541035.1 | 6 | 792 | |||
XM_017017453.1 | XP_016872942.1 | 6 | 948 | |||
XM_017017454.1 | XP_016872943.1 | 6 | 948 | |||
GenBank mRNA | AF230388.1 | - | 6 | 1615 | ||
AF230389.1 | - | 6 | 1615 | |||
AK093404.1 | - | 2 | 685 | |||
AK122720.1 | - | 7 | 1431 | |||
AK223090.1 | - | 6 | 1573 | |||
AK299459.1 | - | 6 | 893 | |||
AK299472.1 | - | 6 | 972 | |||
AK303969.1 | - | 6 | 853 | |||
BC010644.1 | - | 6 | 1561 | 3855016 | ||
BC017352.2 | - | 6 | 1554 | 5013158 | ||
L24203.1 | - | 6 | 1615 |