Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318082.1 | NP_001305011.1 | 7 | 2668 | ||
NM_001318083.1 | NP_001305012.1 | 5 | 2387 | |||
NM_001318084.1 | NP_001305013.1 | 5 | 2380 | |||
NM_014569.3 | NP_055384.1 | 7 | 2727 | |||
NM_145102.3 | NP_659570.1 | 7 | 2606 | |||
XM_011515999.2 | XP_011514301.1 | 6 | 2267 | |||
XM_017011918.1 | XP_016867407.1 | 9 | 3065 | |||
XM_017011919.1 | XP_016867408.1 | 8 | 3108 | |||
XM_017011920.1 | XP_016867409.1 | 6 | 3270 | |||
XM_017011921.1 | XP_016867410.1 | 7 | 3036 | |||
XM_017011922.1 | XP_016867411.1 | 7 | 2695 | |||
XM_017011923.1 | XP_016867412.1 | 7 | 2846 | |||
XM_017011924.1 | XP_016867413.1 | 5 | 2508 | |||
XM_017011925.1 | XP_016867414.1 | 6 | 2212 | |||
XM_017011926.1 | XP_016867415.1 | 6 | 2630 | |||
XM_017011927.1 | XP_016867416.1 | 6 | 2562 | |||
GenBank mRNA | AB023232.1 | - | 7 | 2668 | ||
AF170025.1 | - | 7 | 2580 | |||
AK027714.1 | - | 7 | 2696 | |||
BC030790.2 | - | 7 | 2637 | 4827870 | ||
BX648490.1 | - | 5 | 3133 |