Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318082.1 | NP_001305011.1 | 5 | 907 | ||
NM_014569.3 | NP_055384.1 | 5 | 966 | |||
NM_145102.3 | NP_659570.1 | 5 | 845 | |||
XM_011515999.2 | XP_011514301.1 | 4 | 506 | |||
XM_017011918.1 | XP_016867407.1 | 7 | 1304 | |||
XM_017011919.1 | XP_016867408.1 | 6 | 1347 | |||
XM_017011920.1 | XP_016867409.1 | 4 | 1509 | |||
XM_017011921.1 | XP_016867410.1 | 5 | 1275 | |||
XM_017011922.1 | XP_016867411.1 | 5 | 934 | |||
XM_017011925.1 | XP_016867414.1 | 4 | 451 | |||
GenBank mRNA | AB023232.1 | - | 5 | 907 | ||
AB742318.1 | - | 4 | 396 | |||
AF170025.1 | - | 5 | 819 | |||
AK027714.1 | - | 5 | 935 | |||
AK096552.1 | - | 5 | 1504 | |||
BC030790.2 | - | 5 | 876 | 4827870 |