Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135820.1 | NP_001129292.1 | 10 | 2451 | ||
NM_013390.2 | NP_037522.1 | 11 | 2640 | |||
XM_005251869.4 | XP_005251926.1 | 11 | 2273 | |||
GenBank mRNA | AB037833.2 | - | 11 | 2233 | ||
AF137030.1 | - | 11 | 2248 | |||
AK075370.1 | - | 4 | 368 | |||
BC140773.1 | - | 10 | 1953 | 9021641 | ||
BC144510.1 | - | 11 | 2108 | |||
BC146780.1 | - | 11 | 2233 | 9007233 | ||
BC171782.1 | - | 10 | 1954 |