Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014521.2 | NP_055336.1 | 4 | 2004 | ||
XM_011510891.2 | XP_011509193.1 | 4 | 2024 | |||
XM_011510892.1 | XP_011509194.1 | 4 | 2013 | |||
XM_011510893.1 | XP_011509195.1 | 4 | 1938 | |||
XM_011510894.2 | XP_011509196.1 | 4 | 4437 | |||
GenBank mRNA | AF147747.1 | - | 4 | 1946 | ||
AK291209.1 | - | 4 | 1971 | |||
AK304398.1 | - | 1 | 164 | |||
BC057396.1 | - | 4 | 2015 | 6138465 | ||
BX647198.1 | - | 4 | 1931 | |||
BX648877.1 | - | 4 | 2119 | |||
DQ232895.1 | - | 4 | 1978 |