Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013276.2 | NP_037408.2 | 3 | 573 | ||
GenBank mRNA | AF163573.1 | - | 3 | 573 | ||
AK055609.1 | - | 3 | 499 | |||
AK312428.1 | - | 3 | 520 | |||
AL832420.1 | - | 3 | 512 | |||
BC020543.1 | - | 3 | 529 | 4413491 |