Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032012.3 | NP_114401.2 | 5 | 1053 | ||
XM_011518446.2 | XP_011516748.1 | 5 | 1068 | |||
XM_011518447.1 | XP_011516749.1 | 5 | 1053 | |||
XM_011518449.1 | XP_011516751.1 | 4 | 936 | |||
XM_011518451.1 | XP_011516753.1 | 5 | 1053 | |||
XM_017014571.1 | XP_016870060.1 | 5 | 1050 | |||
GenBank mRNA | AF153415.1 | - | 5 | 1053 | ||
AK299952.1 | - | 5 | 1035 | |||
AL833960.1 | - | 4 | 360 | |||
BP360669.1 | - | 2 | 200 | |||
CX865814.1 | - | 2 | 141 |