Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282738.1 | NP_001269667.1 | 7, 8 | 1252 | ||
NM_001282739.1 | NP_001269668.1 | 6, 7 | 1572 | |||
NM_001282740.1 | NP_001269669.1 | 5, 6 | 755 | |||
NM_001282741.1 | NP_001269670.1 | 5, 6 | 812 | |||
NM_001282742.1 | NP_001269671.1 | 4, 5 | 483 | |||
NM_030758.3 | NP_110385.1 | 6, 7 | 1575 | |||
XM_006724207.2 | XP_006724270.1 | 5, 6 | 758 | |||
XM_006724208.3 | XP_006724271.1 | 4, 5 | 543 | |||
XM_011530059.2 | XP_011528361.1 | 6, 7 | 1501 | |||
XM_011530060.1 | XP_011528362.1 | 5, 6 | 812 | |||
XM_011530062.1 | XP_011528364.1 | 6, 7 | 857 | |||
XM_017028709.1 | XP_016884198.1 | 6, 7 | 1498 | |||
XM_017028710.1 | XP_016884199.1 | 6, 7 | 964 | |||
XM_017028712.1 | XP_016884201.1 | 5, 6 | 759 | |||
XM_017028713.1 | XP_016884202.1 | 5, 6 | 1085 | |||
GenBank mRNA | AB051451.2 | - | 6, 7 | 1483 | ||
AF288741.1 | - | 6, 7 | 1469 | |||
AF323731.1 | - | 6, 7 | 1575 | |||
AK093196.1 | - | 5, 6 | 755 | |||
AK131374.1 | - | 6, 7 | 1102 | |||
AK296354.1 | - | 5, 6 | 812 | |||
AK296525.1 | - | 7, 8 | 1254 | |||
AK300323.1 | - | 4, 5 | 483 | |||
BC004340.1 | - | 3, 4 | 370 | |||
BC118914.2 | - | 6, 7 | 1489 | 40037933 |