Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289905.1 | NP_001276834.1 | 8, 9 | 979 | ||
NM_014339.6 | NP_055154.3 | 8, 9 | 979 | |||
GenBank mRNA | BC011624.2 | - | 8, 9 | 923 | 4053746 | |
BX368715.2 | - | 2, 3 | 150 | |||
BX410574.2 | - | 8, 9 | 896 | |||
CT841520.1 | - | 8, 9 | 902 | |||
U58917.1 | - | 8, 9 | 878 |