Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013231.4 | NP_037363.1 | 2 | 2278 | ||
XM_005267490.3 | XP_005267547.1 | 2 | 2377 | |||
XM_011536611.2 | XP_011534913.1 | 2 | 2018 | |||
XM_017021129.1 | XP_016876618.1 | 2 | 2559 | |||
XM_017021130.1 | XP_016876619.1 | 2 | 11833 | |||
XM_017021131.1 | XP_016876620.1 | 3 | 10987 | |||
XM_017021132.1 | XP_016876621.1 | 3 | 2466 | |||
XM_017021133.1 | XP_016876622.1 | 3 | 2484 | |||
XM_017021134.1 | XP_016876623.1 | 3 | 2411 | |||
XM_017021135.1 | XP_016876624.1 | 2 | 2279 | |||
GenBank mRNA | AF169676.1 | - | 1 | 1568 | ||
AK056649.1 | - | 2 | 2278 | |||
AY358287.1 | - | 1 | 1774 | |||
BC126249.1 | - | 2 | 2194 | 8991965 | ||
BC130290.1 | - | 2 | 2194 | 40146321 | ||
BC143936.1 | - | 2 | 2216 | 9052454 |