Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013280.4 | NP_037412.2 | 2 | 689 | ||
XM_005273861.1 | XP_005273918.1 | 3 | 1575 | |||
XM_011544883.1 | XP_011543185.1 | 2 | 529 | |||
GenBank mRNA | AF169675.1 | - | 1 | 348 | ||
AK289931.1 | - | 2 | 1006 | |||
AY358308.1 | - | 2 | 810 | |||
BC018370.1 | - | 2 | 689 | 4214494 |